Supelco® PESTANAL® Organic Reference Standard, Captan
Catalog # 32054-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:133-06-2
- Description:Captan
- Storage conditions:2...30 °C
- Hazard class description:H317 - H318 - H331 - H351 - H400
- Shipping conditions:STD
- MDL No.:MFCD00041811
- M-Clarity quality level:MQ100
- Formula:C₉H₈Cl₃NO₂S
- Molecular weight:300,59
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS08, GHS05, GHS09
Caution: P273 - P280 - P302 + P352 - P304 + P340 + P311 - P305 + P351 + P338 - P308 + P313