Supelco® PESTANAL® Organic Reference Standard, Butocarboxim
36121-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard, mixture of isomers
- CAS No.:34681-10-2
- Description:Butocarboxim
- Storage conditions:2...30 °C
- Hazard class description:H226 - H301 + H311 + H331 - H319 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055326
- M-Clarity quality level:MQ100
- Formula:C₇H₁₄N₂O₂S
- Molecular weight:190,26
- Flash point:Not applicable
- GHS signal: GHS02, GHS06, GHS09
: P210 - P273 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311