Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Butamifos
Supelco® PESTANAL® Organic Reference Standard, Butamifos
  37046-25MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Butamifos
  37046-25MG
 :  Merck
 :  

 

  • Pk:
    25 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    36335-67-8
  • Description:
    Butamifos
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302 - H410
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00210251
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    O-Ethyl O-(5-methyl-2-nitrophenyl) N-sec-butylphosphorothionoamidate; O-Ethyl O-6-nitro-m-tolyl sec-butylphosphoramidothioate
  • Formula:
    C₁₆H₁₄O₅
  • Molecular weight:
    332,36
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS07, GHS09
 : P264 - P270 - P273 - P301 + P312 - P391 - P501