Supelco® PESTANAL® Organic Reference Standard, Bromadiolon
Catalog # 46035-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:28772-56-7
- Description:Bromadiolon
- Storage conditions:2...30 °C
- Hazard class description:H300 + H310 + H330 - H360D - H372 - H410
- Shipping conditions:STD
- MDL No.:MFCD00128053
- M-Clarity quality level:MQ100
- Synonym(s):Bromodiolone
- Molecular weight:527,41
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P202 - P264 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310