Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, β-Cyfluthrin
Supelco® PESTANAL® Organic Reference Standard, β-Cyfluthrin
  46003-250MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, β-Cyfluthrin
  46003-250MG
 :  Merck
 :  

 

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, analytical standard
  • CAS No.:
    1820573-27-0
  • Description:
    β-Cyfluthrin
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H300 - H331 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00078636
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    434,29

 

 

  • GHS signal: GHS06, GHS09
 : P261 - P264 - P270 - P273 - P301 + P310 - P304 + P340 + P311