Supelco® PESTANAL® Organic Reference Standard, Bensulfuron-Methyl
Catalog # 37897-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:83055-99-6
- Description:Bensulfuron-methyl
- Storage conditions:2...30 °C
- Hazard class description:H317 - H410
- Shipping conditions:STD
- MDL No.:MFCD00128064
- M-Clarity quality level:MQ100
- Melting point:179...184 °C
- Formula:C₁₆H₁₈N₄O₇S
- Molecular weight:410,40
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P261 - P272 - P273 - P280 - P302 + P352 - P333 + P313