Supelco® PESTANAL® Organic Reference Standard, Benomyl
Catalog # 45339-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:17804-35-2
- Description:Benomyl
- Storage conditions:2...8 °C
- Hazard class description:H315 - H317 - H335 - H340 - H360 - H410
- Shipping conditions:STD
- MDL No.:MFCD00084636
- M-Clarity quality level:MQ100
- Synonym(s):Methyl 1-(butylcarbamoyl)-2-benzimidazolecarbamate; Benomyl; NSC 263489
- Melting point:>300 °C (lit.)
- Formula:C₁₄H₁₈N₄O₃
- Molecular weight:290,32
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS08, GHS07, GHS09
Caution: P202 - P261 - P273 - P280 - P302 + P352 - P308 + P313