Supelco® PESTANAL® Organic Reference Standard, Benazolin-Ethyl Ester
31227-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:25059-80-7
- Description:Benazolin-ethyl ester
- Storage conditions:2...30 °C
- Hazard class description:H411
- Shipping conditions:STD
- MDL No.:MFCD00143653
- M-Clarity quality level:MQ100
- Synonym(s):4-Chloro-2-oxo-3(2H)-benzothiazoleacetic acid ethyl ester; Ethyl-4-chlor-2-oxo-benzothiazolinacetat
- Formula:C₁₁H₁₀ClNO₃S
- Molecular weight:271,72
- Flash point:Not applicable
- GHS signal: GHS09
: P273 - P391 - P501