Supelco® Organic Reference Standard, Behenic Acid
Supplier: Merck
Certificates
About this item
Specifications
- CAS No.:112-85-6
- Storage conditions:RT
- Shipping conditions:STD
- MDL No.:MFCD00002807
- M-Clarity quality level:MQ100
- Synonym(s):Docosanoic acid
- Melting point:72...80 °C (lit.); 79...82 °C
- Formula:CH₃(CH₂)₂₀COOH
- Molecular weight:340,58
- Flash point:Not applicable
Specifications
Frequently Bought Together