Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Azamethiphos
Supelco® PESTANAL® Organic Reference Standard, Azamethiphos
Catalog # 45331-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Azamethiphos
Catalog # 45331-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    35575-96-3
  • Description:
    Azamethiphos
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302 - H317 - H331 - H351 - H370 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00867611
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    S-((6-Chloro-2-oxooxazolo[4,5-b]pyridin-3(2H)-yl)methyl) O,O-dimethyl phosphorothioate
  • Formula:
    C₉H₁₀ClN₂O₅PS
  • Molecular weight:
    324,68
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P273 - P280 - P301 + P312 - P302 + P352 - P304 + P340 + P311 - P308 + P311