Supelco® PESTANAL® Organic Reference Standard, Aminocarb
Catalog # 45322-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:2032-59-9
- Description:Aminocarb
- Storage conditions:2...30 °C
- Hazard class description:H300 - H311 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055440
- M-Clarity quality level:MQ100
- Synonym(s):(4-Dimethylamino-3-methylphenyl) N-methylcarbamate
- Formula:C₁₁H₁₆N₂O₂
- Molecular weight:208,26
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P264 - P270 - P273 - P280 - P301 + P310 - P302 + P352 + P312