Supelco® PESTANAL® Organic Reference Standard, α-HCH-d₆
Catalog # 03811-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:86194-41-4
- Description:α-HCH-d₆
- Storage conditions:2...8 °C
- Hazard class description:H301 - H312 - H351 - H410
- Shipping conditions:STD
- MDL No.:MFCD01632293
- M-Clarity quality level:MQ100
- Synonym(s):α-1,2,3,4,5,6-Hexachlorocyclohexane-d₆; α-BHC-d₆
- Melting point:159...160 °C
- Formula:C₆D₆Cl₆
- Molecular weight:296,87
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS08, GHS09
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Caution: P202 - P264 - P273 - P280 - P301 + P310 - P302 + P352 + P312