Supelco® PESTANAL® Organic Reference Standard, Aldicarb-Sulfone
Catalog # 33387-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:1646-88-4
- Description:Aldicarb-sulfone
- Storage conditions:2...30 °C
- Hazard class description:H300 + H310 + H330 - H400
- Shipping conditions:STD
- MDL No.:MFCD00041804
- M-Clarity quality level:MQ100
- Melting point:132...135 °C
- Formula:C₇H₁₄N₂O₄S
- Molecular weight:222,26
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P260 - P262 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310