Supelco® PESTANAL® Organic Reference Standard, Alachlor ESA Sodium Salt
Catalog # 34147-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:140939-15-7
- Description:Alachlor ESA sodium salt
- Storage conditions:2...30 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD09037344
- M-Clarity quality level:MQ100
- Synonym(s):2-[(2,6-Diethylphenyl)(methoxymethyl)-amino]-2-oxo-ethanesulfonic acid sodium salt
- Formula:C₁₄H₂₀NNaO₅S
- Molecular weight:337,37
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338