Order Entry
Slovakia
ContactUsLinkComponent
Organic reference standard, 2-Methyl-4-isothiazolin-3-one, analytical standard, Supelco®
Organic reference standard, 2-Methyl-4-isothiazolin-3-one, analytical standard, Supelco®
  73569-1G
 :  Merck
 :  
undefined
Organic reference standard, 2-Methyl-4-isothiazolin-3-one, analytical standard, Supelco®
  73569-1G
 :  Merck
 :  

 

  • Pk:
    1 g
  • Standards:
    Analytical standard
  • CAS No.:
    2682-20-4
  • Description:
    2-Methyl-4-isothiazolin-3-one
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₄H₅NOS
  • Description:
    2-Methyl-4-isothiazolin-3-one