Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, 2-Dimethylaminoethanol
Supelco® Organic Reference Standard, 2-Dimethylaminoethanol
Catalog # 50632-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 2-Dimethylaminoethanol
Catalog # 50632-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    108-01-0
  • Description:
    2-Dimethylaminoethanol
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H226 - H302 + H312 - H314 - H331 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00002846
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    N,N-Dimethyl-2-hydroxyethylamine; N,N-Dimethylethanolamine; DMEA; Jeffcat DMEA
  • Melting point:
    –70 °C (lit.)
  • Formula:
    (CH₃)₂NCH₂CH₂OH
  • Molecular weight:
    89,14
  • Density:
    0,886 g/ml at 20 °C (lit.)
  • Flash point:
    40 °C - Seta Closed cup
  • Boiling point:
    134 - 136 °C (lit.)

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS05
Caution: P210 - P280 - P301 + P312 + P330 - P303 + P361 + P353 - P304 + P340 + P311 - P305 + P351 + P338 + P310