Supelco® Organic Reference Standard, Unleaded Gasoline
UST155-1.5ML
: Merck
:
- Concentration:5000 µg/ml in methanol
- Pk:1,5 mL
- Standards:Certified reference material
- Description:Unleaded gasoline
- Storage conditions:2...30 °C
- Hazard class description:H225 - H301 + H311 + H331 - H340 - H350 - H370
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:11 °C - Closed cup - Solvent
- GHS signal: GHS02, GHS06, GHS08
: P202 - P210 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311