Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Aliphatics Mix (C5-C12)
  UST157-1.5ML
 :  Merck
 :  
Supelco® Organic Reference Standard, Aliphatics Mix (C5-C12)
  UST157-1.5ML
 :  Merck
 :  

 

  • Concentration:
    2000 µg/ml each component in methanol
  • Pk:
    1,5 mL
  • Standards:
    Certified reference material
  • Description:
    Aliphatics mix (C5-C12)
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370 - H412
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Flash point:
    11 °C - Closed cup - Solvent

 

 

  • GHS signal: GHS02, GHS06, GHS08
 : P210 - P273 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311