Chlorite ion chromatography (IC) standard, Supelco®
Supplier: Merck
Certificates
About this item
Chlorite IC standard may be used in the determination of disinfection by-product, chlorite in water samples using ion chromatography technique (IC)
Certifications: his certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Frequently Bought Together