Organic reference standard, 2-isopropyl-6-methyl-4-pyrimidinol, Dr Ehrenstorfer
Catalog # EHERC14463800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:0,25 g
- Standards:Reference standard
- CAS No.:2814-20-2
- Description:2-Isopropyl-6-methyl-4-pyrimidinol
- Formula:C₈H₁₂N₂O
- Description:2-Isopropyl-6-methyl-4-pyrimidinol