Supelco® Organic Reference Standard, Lead - Soil
Catalog # PB3000-50G
Supplier: Merck
CAS Number:
Specifications
- Concentration:Pk of 50 g
- Pk:50 g
- Standards:Certified reference material
- Description:Lead - soil
- Storage conditions:2...30 °C
- Hazard class description:H360FD - H412
- Shipping conditions:STD
Specifications
About this item
- GHS signal: GHS08, GHS09
Caution: P202 - P273 - P280 - P308 + P313 - P405 - P501