Order Entry
Slovakia
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, EPA 501/601 Trihalomethanes Calibration Mix
Supelco® TraceCERT® Organic Reference Standard, EPA 501/601 Trihalomethanes Calibration Mix
Supplier:  Merck
Certificates
Supelco® TraceCERT® Organic Reference Standard, EPA 501/601 Trihalomethanes Calibration Mix
Supplier:  Merck

Specifications

  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ100
  • Flash point:
    9,7 °C - Closed cup

Specifications

Frequently Bought Together