Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Anions - Loamy sand 1, (CRM), Supelco®
Anions - Loamy sand 1, (CRM), Supelco®
Anions - Loamy sand 1, (CRM), Supelco®
Catalog # CRM702-50G
Supplier:
Merck
CAS Number:
Expand image
Anions - Loamy sand 1, (CRM), Supelco®
Catalog # CRM702-50G
Supplier:
Merck
CAS Number:
Specifications
Pk:
50 g
Description:
Anions - Loamy Sand 1
Pk Info:
Pkg of 50 g
Type:
Certified reference material
Specifications
About this item
Product Details
Product Details