Supelco® Organic Reference Standard, p-Cymene solution
CRM40419
: Merck
:
- Concentration:2000 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:99-87-6
- Description:p-Cymene solution
- Storage conditions:2…8 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:WICE
- M-Clarity quality level:MQ300
- Synonym(s):p-Isopropyltoluene
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
- GHS signal: GHS02, GHS06, GHS08
: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311