Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Longifolene solution
Supelco® Organic Reference Standard, Longifolene solution
Catalog # CRM40481
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Longifolene solution
Catalog # CRM40481
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    475-20-7
  • Description:
    Longifolene solution
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370 - H412
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ300
  • Flash point:
    9,7 °C - Closed cup
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P273 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311