Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, alpha-Humulene solution
  CRM40921
 :  Merck
 :  
Supelco® Organic Reference Standard, alpha-Humulene solution
  CRM40921
 :  Merck
 :  

 

  • Concentration:
    2000 µg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    6753-98-6
  • Description:
    alpha-Humulene solution
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ300
  • Flash point:
    9,7 °C - Closed cup
  • Pk Info:
    Ampule of 1 ml

 

 

  • GHS signal: GHS02, GHS06, GHS08
 : P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311