0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovakia
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical secondary standard, risperidone, (CRM), Supelco®
Pharmaceutical secondary standard, risperidone, (CRM), Supelco®
PHR1631-500MG
:
Merck
:
Pharmaceutical secondary standard, risperidone, (CRM), Supelco®
PHR1631-500MG
:
Merck
:
Pk:
500 mg
CAS No.:
106266-06-2
Description:
Risperidone
Description:
Risperidone