0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovakia
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical secondary standard, m-Xylene, (CRM), Supelco®
Pharmaceutical secondary standard, m-Xylene, (CRM), Supelco®
Catalog # PHR1313-1G
Supplier:
Merck
CAS Number:
Expand image
Pharmaceutical secondary standard, m-Xylene, (CRM), Supelco®
Catalog # PHR1313-1G
Supplier:
Merck
CAS Number:
Specifications
Pk:
1 g
CAS No.:
108-38-3
Description:
m-Xylene
Description:
m-Xylene
Specifications
About this item
Product Details
Product Details