Supelco® Organic Reference Standard, Lidocaine Impurity D
PHR1977-50MG
: Merck
:
- Pk:50 mg
- Standards:Pharmaceutical Secondary Standard; Certified Reference Material
- Description:Lidocaine Impurity D
- Seq:25
- Storage conditions:2...30 °C
- Hazard class description:H302 - H315 - H319
- Shipping conditions:STD
- M-Clarity quality level:MQ300
- Synonym(s):NORLIDOCAINE HYDROCHLORIDE
- GHS signal: GHS07
: P301 + P312 + P330 - P305 + P351 + P338