Pharmaceutical secondary standard, Lidocaine and related compounds, (CRM), Supelco®
: Merck
Lidocaine is a drug, which serves as a potential candidate in the treatment of cardiac arrhythmias.
Lidocaine is a local anesthetic drug that can exhibit pronounced antiarrhythmic and anticonvulsant effects. It is known as a central nervous system depressant and shows sedative, analgesic and anticonvulsant properties.
: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025
- CAS No.:137-58-6
- Formula:C₁₄H₂₂N₂O
- Description:Lidocaine
Lidocaine Impurities
Frequently Bought Together