Pharmaceutical secondary standard, Homosalate, (CRM), Supelco®
: Merck
- CAS No.:118-56-9
- Description:Homosalate
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Safety wash bottles, wide neck, multilingual, Azlon®
: DWK LIFE SCIENCES
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Sodium iodide, anhydrous 99+%
: Apollo Scientific