Order Entry
Slovakia
ContactUsLinkComponent
Pharmaceutical secondary standard, cholecalciferol (Vitamin D3), (CRM), Supelco®
Pharmaceutical secondary standard, cholecalciferol (Vitamin D3), (CRM), Supelco®
Catalog # PHR1237-500MG
Supplier:  Merck
CAS Number:  
Pharmaceutical secondary standard, cholecalciferol (Vitamin D3), (CRM), Supelco®
Catalog # PHR1237-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 mg
  • CAS No.:
    67-97-0
  • Description:
    Cholecalciferol (Vitamin D3)
  • Melting point:
    83…86 °C (lit.)
  • Formula:
    C₂₇H₄₄O
  • Description:
    Cholecalciferol (Vitamin D3)
  • Form:
    Neat

Specifications

About this item

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

  • Traceable to BP 787
  • Traceable to PhEur C2100000
  • Traceable to USP 1131009

Vitamin D acts through a receptor that is a member of the ligand-dependent transcription factor superfamily. Modulates the proliferation and differentiation of both normal and cancer cells. Has antiproliferative and antimetastatic effects on breast, colon, and prostate cancer cells. Activated vitamin D receptors in intestine and bone maintain calcium absorbance and homeostasis.

Certifications: This product was designed, produced and verified for accuracy and stability in accordance with ISO/IEC 17025:2005 (AClass Cert AT-1467), ISO GUIDES 34:2009 (AClass Cert AR-1470).