0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovakia
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical secondary standard, Cefaclor, (CRM), Supelco®
Pharmaceutical secondary standard, Cefaclor, (CRM), Supelco®
PHR1283-1G
:
Merck
:
Pharmaceutical secondary standard, Cefaclor, (CRM), Supelco®
PHR1283-1G
:
Merck
:
Pk:
1 g
CAS No.:
70356-03-5
Description:
Cefaclor
Description:
Cefaclor