Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Acetaminophen Impurity I
Supelco® Organic Reference Standard, Acetaminophen Impurity I
Catalog # PHR1973-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Acetaminophen Impurity I
Catalog # PHR1973-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Pharmaceutical Secondary Standards; Certified Reference Material
  • CAS No.:
    118-93-4
  • Description:
    Acetaminophen impurity I
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00002219
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    2′-Hydroxyacetophenone; 2′-Hydroxyacetophenone; 2-Acetylphenol
  • Melting point:
    3 - 6 °C (lit.)
  • Formula:
    HOC₆H₄COCH₃
  • Molecular weight:
    136,15
  • Density:
    1,133 g/ml at 20 °C; 1,131 g/ml at 25 °C (lit.)
  • Flash point:
    106 °C - Closed cup
  • Boiling point:
    213 °C/717 mmHg (lit.)

Specifications

About this item