Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® TraceCERT® Organic Reference Standard, Salicylic acid
Catalog # 52341-50MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® TraceCERT® Organic Reference Standard, Salicylic acid
Catalog # 52341-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    69-72-7
  • Description:
    Salicylic acid
  • Storage conditions:
    15...25 °C
  • Hazard class description:
    H302 - H318 - H361d
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00002439
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    2-Hydroxybenzoic acid
  • Melting point:
    158 - 161 °C (lit.)
  • Formula:
    2⁻(HO)C₆H₄CO₂H
  • Molecular weight:
    138,12
  • Flash point:
    157 °C - Closed cup
  • Boiling point:
    211 °C (lit.)

Specifications

About this item

  • GHS signal: GHS05, GHS07
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P202 - P264 - P280 - P301 + P312 - P305 + P351 + P338 - P308 + P313