Supelco® TraceCERT® Organic Reference Standard, ʟ-Serine
Katalogové číslo 54763-100MG
Dodavatel: Merck
Číslo CAS:
Specifikace
- Pk:100 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:56-45-1
- Description:ʟ-Serine
- Storage conditions:15...25 °C
- Shipping conditions:STD
- MDL No.:MFCD00064224
- M-Clarity quality level:MQ300
- Synonym(s):(S)-2-Amino-3-hydroxypropionic acid
- Melting point:222 °C (dec.) (lit.)
- Formula:HOCH₂CH(NH₂)CO₂H
- Molecular weight:105,09
Specifikace
O tomto produktu
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.