Order Entry
Slovakia
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, ʟ-Lysine monohydrochloride
Supelco® TraceCERT® Organic Reference Standard, ʟ-Lysine monohydrochloride
  67448-100MG
 :  Merck
 :  
undefined
Supelco® TraceCERT® Organic Reference Standard, ʟ-Lysine monohydrochloride
  67448-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    657-27-2
  • Description:
    ʟ-Lysine monohydrochloride
  • Storage conditions:
    15...25 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00064564
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    ⁽S⁾-2,6-Diaminohexanoic acid monohydrochloride
  • Melting point:
    263 °C (dec.) (lit.)
  • Formula:
    H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
  • Molecular weight:
    182,65

 

 

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.