Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® TraceCERT® Organic Reference Standard, ʟ-Glutamine
  76523-100MG
 :  Merck
 :  
undefined
Supelco® TraceCERT® Organic Reference Standard, ʟ-Glutamine
  76523-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    56-85-9
  • Description:
    ʟ-Glutamine
  • Storage conditions:
    15...25 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00008044
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    (S)-2,5-Diamino-5-oxopentanoic acid; ʟ-Glutamic acid 5-amide; Levoglutamide
  • Melting point:
    185 °C (dec.) (lit.)
  • Formula:
    H₂NCOCH₂CH₂CH(NH₂)CO₂H
  • Molecular weight:
    146,14

 

 

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.