Supelco® TraceCERT® Organic Reference Standard, ʟ-Arginine monohydrochloride
90538-100MG
: Merck
:
- Pk:100 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:1119-34-2
- Description:ʟ-Arginine monohydrochloride
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD00064550
- M-Clarity quality level:MQ300
- Synonym(s):(S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride; (S)-(+)-Arginine hydrochloride
- Melting point:226 - 230 °C
- Molecular weight:210,66
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.