Order Entry
Slovakia
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Hydroquinone
Supelco® TraceCERT® Organic Reference Standard, Hydroquinone
Katalogové číslo 74347-100MG
Dodavatel:  Merck
Číslo CAS:  
undefined
Supelco® TraceCERT® Organic Reference Standard, Hydroquinone
Katalogové číslo 74347-100MG
Dodavatel:  Merck
Číslo CAS:  

Specifikace

  • Pk:
    100 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    123-31-9
  • Description:
    Hydroquinone
  • Storage conditions:
    15...25 °C
  • Hazard class description:
    H302 - H317 - H318 - H341 - H351 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00002339
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    1,4-Benzenediol; 1,4-Dihydroxybenzene; HQ
  • Melting point:
    172 - 175 °C (lit.)
  • Formula:
    C₆H₄-1,4-(OH)₂
  • Molecular weight:
    110,11
  • Flash point:
    165 °C - Closed cup
  • Boiling point:
    285 °C (lit.)

Specifikace

O tomto produktu

  • GHS signal: GHS08, GHS05, GHS07, GHS09
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P273 - P280 - P301 + P312 - P302 + P352 - P305 + P351 + P338 - P308 + P313