Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Anthracene
Supelco® Organic Reference Standard, Anthracene
  07671-100MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Anthracene
  07671-100MG
 :  Merck
 :  

 

  • Pk:
    100 mL
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    120-12-7
  • Description:
    Anthracene
  • Storage conditions:
    15...25 °C
  • Hazard class description:
    H319 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00001240
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    Anthraxcene; Paranaphthalene
  • Melting point:
    210...215 °C (lit.)
  • Molecular weight:
    178,23
  • Flash point:
    121,0 °C - closed cup
  • Boiling point:
    340 °C (lit.)

 

 

  • GHS signal: GHS09
 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
 : P264 - P273 - P280 - P305 + P351 + P338 - P337 + P313 - P391