Supelco® Drug and metabolite standards, O-desmethyl-cis-tramadol-D6 hydrochloride solution, (CRM)
D-058-1ML
: Merck
:
- Concentration:100 µg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:126193-87-6
- Description:O-Desmethyl-cis-tramadol-D6 hydrochloride solution
- Environmentally Preferable:
- Description:O-Desmethyl-cis-tramadol-D6 hydrochloride solution
- Pk Info:ampoule of 1 ml