Supelco® Cerilliant® Organic Reference Standard, Estrone solution
Catalog # E-075-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:53-16-7
- Description:Estrone solution
- Storage conditions:–20 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:STD
- MDL No.:MFCD00003620
- M-Clarity quality level:MQ300
- Molecular weight:270,37
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311