Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Cerilliant® Organic Reference Standard, Dopamine hydrochloride solution
Catalog # D-081-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Cerilliant® Organic Reference Standard, Dopamine hydrochloride solution
Catalog # D-081-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1,0 mg/ml in methanol with 5% 1 M HCl (as free base)
  • Pk:
    1 mL
  • Standards:
    Certified reference material, Cerilliant®
  • CAS No.:
    62-31-7
  • Description:
    Dopamine hydrochloride solution
  • Storage conditions:
    –70 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    189,64
  • Flash point:
    11 °C - Closed cup
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311