Supelco® Drug and metabolite standards, Demoxepam solution, (CRM)
Catalog # D-079-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:963-39-3
- Description:Demoxepam solution
- Description:Demoxepam solution
- Pk Info:ampoule of 1 ml