Supelco® Drug and metabolite standards, 3-Trifluoromethylphenylpiperazine (TFMPP)-D4 hydrochloride solution, (CRM)
Catalog # T-920-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:15532-75-9
- Description:3-Trifluoromethylphenylpiperazine (TFMPP)-D4 hydrochloride solution
- Description:3-Trifluoromethylphenylpiperazine (TFMPP)-D4 hydrochloride solution
- Pk Info:ampoule of 1 ml