Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Cerilliant® Organic Reference Standard, 25-Hydroxyvitamin D3 solution
Supelco® Cerilliant® Organic Reference Standard, 25-Hydroxyvitamin D3 solution
Catalog # H-083-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Cerilliant® Organic Reference Standard, 25-Hydroxyvitamin D3 solution
Catalog # H-083-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in ethanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material, Cerilliant®
  • CAS No.:
    63283-36-3
  • Description:
    25-Hydroxyvitamin D3 solution
  • Storage conditions:
    –70 °C
  • Hazard class description:
    H225 - H319
  • Shipping conditions:
    DICE
  • MDL No.:
    MFCD00867077
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    400,64
  • Flash point:
    14 °C - Closed cup
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P233 - P240 - P241 - P242 - P305 + P351 + P338