Supelco® Cerilliant® Organic Reference Standard, (±)-Norepinephrine-D₆ hydrochloride solution
N-069-1ML
: Merck
:
- Concentration:100 µg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:1219803-04-9
- Description:(±)-Norepinephrine-D₆ hydrochloride solution
- Storage conditions:–70 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:DICE
- M-Clarity quality level:MQ300
- Molecular weight:211,68
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
- GHS signal: GHS02, GHS06, GHS08
: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311