Supelco® Cerilliant® Organic Reference Standard, (±)-Epinephrine-D₆ solution
Catalog # E-077-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol with 5% 1 M HCl
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:1219803-77-6
- Description:(±)-Epinephrine-D₆ solution
- Storage conditions:–70 °C
- Hazard class description:H225 - H290 - H301 + H311 + H331 - H370
- Shipping conditions:DICE
- M-Clarity quality level:MQ300
- Synonym(s):(±)-Epinephrine-D₆
- Molecular weight:189,24
- Flash point:11 °C - Closed cup
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311