Supelco® Organic Reference Standard, Grain FAME Mix
CRM47801
: Merck
:
- Concentration:In dichloromethane
- Pk:1
- Standards:Certified reference material
- Description:Grain FAME mix
- Storage conditions:–10...–25 °C
- Hazard class description:H315 - H319 - H336 - H351
- Shipping conditions:DICE
- M-Clarity quality level:MQ100
- Pk Info:Ampule of 1 ml
- GHS signal: GHS08, GHS07
: P201 - P202 - P261 - P302 + P352 - P305 + P351 + P338 - P308 + P313